Learn More
Chemglass Life Sciences 60mL Separatory Funnel, Squibb, #16 Outer Stopper Neck, 2mm PTFE Stpk
SDP
Catalog No. CG174202
Encompass_Preferred
60mL Separatory Funnel, Squibb, #16 Outer Stopper Neck, 2mm PTFE Stpk. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.
Catalog No. CG174202 Supplier Chemglass Life Sciences Supplier No. CG174202
60mL Separatory Funnel, Squibb, #16 Outer Stopper Neck, 2mm PTFE Stpk. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.