Learn More
Chemglass Life Sciences 1000mL Separatory Funnel, Squibb, 24/40 Joint Size, 4mm Stpk
SDP
Catalog No. CG175006
Encompass_Preferred
1000mL Separatory Funnel, Squibb, 24/40 Joint Size, 4mm Stpk. Similar to CG-1742, but with the plain lower stem replaced by an inner standard taper joint of the listed size having a sealed in drip tip. Supplied complete with one listed size flask length stopper.
Catalog No. CG175006 Supplier Chemglass Life Sciences Supplier No. CG175006
1000mL Separatory Funnel, Squibb, 24/40 Joint Size, 4mm Stpk. Similar to CG-1742, but with the plain lower stem replaced by an inner standard taper joint of the listed size having a sealed in drip tip. Supplied complete with one listed size flask length stopper.