Learn More
Ace Glass, Inc. Funnel, separatory, PTFE stopcock, 125mL, 19/22
SDP
Catalog No. 50976962
Encompass_Preferred
125ml separatory funnel features a PTFE stopcock and a 19/22 taper joint with drip tip. Stopper is not included.
Catalog No. 50-976-962 Supplier Ace Glass, Inc. Supplier No. 950018
125ml separatory funnel features a PTFE stopcock and a 19/22 taper joint with drip tip. Stopper is not included.