Learn More
Chemglass Life Sciences 2000mL Separatory Funnel, Squibb, #38 Outer Stopper Neck, 6mm Glass Stpk
SDP
Catalog No. 50873280
Encompass_Preferred
2000mL Separatory Funnel, Squibb, #38 Outer Stopper Neck, 6mm Glass Stpk. Squibb style separatory funnel with a glass stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.
Catalog No. 50-873-280 Supplier Chemglass Life Sciences Supplier No. CG174007
2000mL Separatory Funnel, Squibb, #38 Outer Stopper Neck, 6mm Glass Stpk. Squibb style separatory funnel with a glass stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.