Learn More
Chemglass Life Sciences Funnel, Separatory, 4000mL, Squibb, 10mm PTFE Stpk, #38 PTFE Stopper
SDP
Catalog No. 501311094
Encompass_Preferred
Funnel, Separatory, 4000mL, Squibb, 10mm PTFE Stpk, #38 PTFE Stopper. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length PTFE stopper. See CG-3020-A for replacement stoppers.
Catalog No. 50-131-1094 Supplier Chemglass Life Sciences Supplier No. CG1742T09
Funnel, Separatory, 4000mL, Squibb, 10mm PTFE Stpk, #38 PTFE Stopper. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length PTFE stopper. See CG-3020-A for replacement stoppers.