Learn More
Ace Glass, Inc. Funnel, separatory, glass stopcock, PTFE stopper, 4000mL
SDP
Catalog No. 50974681
Encompass_Preferred
4000mL Squibb separatory funnel, with a glass stopcock and a PTFE stopper included.
Catalog No. 50-974-681 Supplier Ace Glass, Inc. Supplier No. 722620
4000mL Squibb separatory funnel, with a glass stopcock and a PTFE stopper included.