Learn More
Hach Company Funnel, Separatory, Glass, 500 mL
Catalog No. 501994358 Shop All Hach Company Products
Encompass
Pear-shaped (Squibb) separatory funnel with PTFE stopcock. Clear glass body allows for easy detection of layers between immiscible liquids, all the way to the stopcock. Highly chemical resistant and fully autoclavable. High density linear polyethylene stopper with closed bottom included.
Catalog No. 50-199-4358 Supplier Hach Company Supplier No. 52049
Pear-shaped (Squibb) separatory funnel with PTFE stopcock. Clear glass body allows for easy detection of layers between immiscible liquids, all the way to the stopcock. Highly chemical resistant and fully autoclavable. High density linear polyethylene stopper with closed bottom included.