Learn More
Chemglass Life Sciences 50mL Cylindrical Separatory Funnel, Graduated, 14/20 Joint Size, 2mm Stpk, 225mm OAH
SDP
Catalog No. CG173414
Encompass_Preferred
50mL Cylindrical Separatory Funnel, Graduated, 14/20 Joint Size, 2mm Stpk, 225mm OAH. Cylindrical shaped separatory funnel having a lower standard taper inner joint and a top outer joint, both being the same size. Graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG173414 Supplier Chemglass Life Sciences Supplier No. CG173414
50mL Cylindrical Separatory Funnel, Graduated, 14/20 Joint Size, 2mm Stpk, 225mm OAH. Cylindrical shaped separatory funnel having a lower standard taper inner joint and a top outer joint, both being the same size. Graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.