Learn More
Chemglass Life Sciences Funnel, Separatory, 6000mL, Squibb, 10mm PTFE Stpk, #38 PTFE Stopper
SDP
Catalog No. 502733579
Encompass_Preferred
A 6,000 mL Squibb-style separatory funnel designed for liquid-liquid extractions and phase separations. It features a 10 mm PTFE stopcock and is supplied with a flask-length #38 stopper for secure sealing.
  • Capacity for large-scale separations (6,000 mL).
  • Squibb-style body for controlled, easy phase transfer.
  • 10 mm PTFE stopcock offers chemical resistance and smooth operation.
  • Supplied with a flask-length stopper for leak-resistant sealing.
  • Suitable for routine laboratory liquid-liquid extractions.
  • Compatible with standard #38 outer stopper neck fittings.
Catalog No. 50-273-3579 Supplier Chemglass Life Sciences Supplier No. CG1742T10
A 6,000 mL Squibb-style separatory funnel designed for liquid-liquid extractions and phase separations. It features a 10 mm PTFE stopcock and is supplied with a flask-length #38 stopper for secure sealing.
  • Capacity for large-scale separations (6,000 mL).
  • Squibb-style body for controlled, easy phase transfer.
  • 10 mm PTFE stopcock offers chemical resistance and smooth operation.
  • Supplied with a flask-length stopper for leak-resistant sealing.
  • Suitable for routine laboratory liquid-liquid extractions.
  • Compatible with standard #38 outer stopper neck fittings.