Learn More
Chemglass Life Sciences 60mL Separatory Funnel, Squibb, 24/40 Joint Size, 2mm Stpk
SDP
Catalog No. 501531224
Encompass_Preferred
60mL Separatory Funnel, Squibb, 24/40 Joint Size, 2mm Stpk. Similar to CG-1742, but with the plain lower stem replaced by an inner standard taper joint of the listed size having a sealed in drip tip. Supplied complete with one listed size flask length stopper.
Catalog No. 50-153-1224 Supplier Chemglass Life Sciences Supplier No. CG175001
60mL Separatory Funnel, Squibb, 24/40 Joint Size, 2mm Stpk. Similar to CG-1742, but with the plain lower stem replaced by an inner standard taper joint of the listed size having a sealed in drip tip. Supplied complete with one listed size flask length stopper.