Learn More
Chemglass Life Sciences Funnel, Separatory, 60mL, Squibb, 2mm PTFE Stpk, #16 PTFE Stopper
SDP
Catalog No. 501311087
Encompass_Preferred
Funnel, Separatory, 60mL, Squibb, 2mm PTFE Stpk, #16 PTFE Stopper. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length PTFE stopper. See CG-3020-A for replacement stoppers.
Catalog No. 50-131-1087 Supplier Chemglass Life Sciences Supplier No. CG1742T02
Funnel, Separatory, 60mL, Squibb, 2mm PTFE Stpk, #16 PTFE Stopper. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length PTFE stopper. See CG-3020-A for replacement stoppers.