Learn More
Chemglass Life Sciences 125mL Cylindrical Separatory Funnel, Graduated, #22 Outer Stopper Neck, 2mm Stpk, 295mm OAH
SDP
Catalog No. CG173002
Encompass_Preferred
125mL Cylindrical Separatory Funnel, Graduated, #22 Outer Stopper Neck, 2mm Stpk, 295mm OAH. Similar to CG-1728, but graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. Supplied complete with listed size stopper. For replacement stoppers see CG-3018. Dimensions are the same as CG-1726.
Catalog No. CG173002 Supplier Chemglass Life Sciences Supplier No. CG173002
125mL Cylindrical Separatory Funnel, Graduated, #22 Outer Stopper Neck, 2mm Stpk, 295mm OAH. Similar to CG-1728, but graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. Supplied complete with listed size stopper. For replacement stoppers see CG-3018. Dimensions are the same as CG-1726.