Learn More
Chemglass Life Sciences 60mL Cylindrical Separatory Funnel, Graduated, #16 Outer Stopper Neck, 2mm Stpk, 235mm OAH
SDP
Catalog No. CG173001
Encompass_Preferred
60mL Cylindrical Separatory Funnel, Graduated, #16 Outer Stopper Neck, 2mm Stpk, 235mm OAH. Similar to CG-1728, but graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. Supplied complete with listed size stopper. For replacement stoppers see CG-3018. Dimensions are the same as CG-1726.
Catalog No. CG173001 Supplier Chemglass Life Sciences Supplier No. CG173001
60mL Cylindrical Separatory Funnel, Graduated, #16 Outer Stopper Neck, 2mm Stpk, 235mm OAH. Similar to CG-1728, but graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. Supplied complete with listed size stopper. For replacement stoppers see CG-3018. Dimensions are the same as CG-1726.