Learn More
Chemglass Life Sciences 2L Globe Shaped Separatory Funnel, Large Capacity, #38 Outer Stopper Neck, 6mm PTFE Stpk
SDP
Catalog No. CG173902
Encompass_Preferred
2L Globe Shaped Separatory Funnel, Large Capacity, #38 Outer Stopper Neck, 6mm PTFE Stpk. Large capacity globe shaped separatory funnel with a choice of glass or PTFE stopcock. Stem length is approx. 130mm below the stopcock on all sizes. For replacement stoppers see CG-3018 for #38, CG-3000 for 45/50.
Catalog No. CG173902 Supplier Chemglass Life Sciences Supplier No. CG173902
2L Globe Shaped Separatory Funnel, Large Capacity, #38 Outer Stopper Neck, 6mm PTFE Stpk. Large capacity globe shaped separatory funnel with a choice of glass or PTFE stopcock. Stem length is approx. 130mm below the stopcock on all sizes. For replacement stoppers see CG-3018 for #38, CG-3000 for 45/50.