Learn More
Chemglass Life Sciences Funnel, Solvent Addition, Stem 12mm OD X 75mm Long
SDP
Catalog No. CG175801
Encompass_Preferred
Funnel, Solvent Addition, Stem 12mm OD X 75mm Long. Design of funnel permits addition of solvents to flash chromatography columns without disturbing the silica-gel / packing layer. Lower end of the funnel is closed and the stem has a series of small holes through which the solvent drains. Top of the funnel is 75mm (3in) in diameter with the stem size as listed.
Catalog No. CG175801 Supplier Chemglass Life Sciences Supplier No. CG175801
Funnel, Solvent Addition, Stem 12mm OD X 75mm Long. Design of funnel permits addition of solvents to flash chromatography columns without disturbing the silica-gel / packing layer. Lower end of the funnel is closed and the stem has a series of small holes through which the solvent drains. Top of the funnel is 75mm (3in) in diameter with the stem size as listed.