Learn More
Chemglass Life Sciences Funnel, Solvent Addition For Chromatography Column, Assorted Pack
SDP
Catalog No. CG1758P01
Encompass_Preferred
Funnel, Solvent Addition For Chromatography Column, Assorted Pack. Unbreakable, polypropylene solvent addition funnels useful for chromatography columns. The unique closed-end design facilitates gentle addition of solvent to the column. Perforated funnel stem permits solvent to pour down the column wall without disturbing the top layer of silica gel. Convenient assortment pack consists of five funnels for use with various column sizes. Approximate dimensions are provided.
Catalog No. CG1758P01 Supplier Chemglass Life Sciences Supplier No. CG1758P01
Funnel, Solvent Addition For Chromatography Column, Assorted Pack. Unbreakable, polypropylene solvent addition funnels useful for chromatography columns. The unique closed-end design facilitates gentle addition of solvent to the column. Perforated funnel stem permits solvent to pour down the column wall without disturbing the top layer of silica gel. Convenient assortment pack consists of five funnels for use with various column sizes. Approximate dimensions are provided.