Learn More
Chemglass Life Sciences Funnel, Vial, PTFE, 1.0" OD, 13-425 GPI Thread
SDP
Catalog No. 50872134
Encompass_Preferred
Funnel, Vial, PTFE, 1.0" OD, 13-425 GPI Thread. Funnel machined from PTFE screws securely onto any standard laboratory vial having a GPI thread. Design allows for easy, safe and mess free transfer of product.PLEASE NOTE: VIALS ARE NOT INCLUDED WITH FUNNELS.
Catalog No. 50-872-134 Supplier Chemglass Life Sciences Supplier No. CG176103
Funnel, Vial, PTFE, 1.0" OD, 13-425 GPI Thread. Funnel machined from PTFE screws securely onto any standard laboratory vial having a GPI thread. Design allows for easy, safe and mess free transfer of product.PLEASE NOTE: VIALS ARE NOT INCLUDED WITH FUNNELS.