Learn More
Chemglass Life Sciences Funnel, Vial, PTFE, 1.4" OD, 18-400 GPI Thread
SDP
Catalog No. 501531231
Encompass_Preferred
Funnel, Vial, PTFE, 1.4" OD, 18-400 GPI Thread. Funnel machined from PTFE screws securely onto any standard laboratory vial having a GPI thread. Design allows for easy, safe and mess free transfer of product.
Catalog No. 50-153-1231 Supplier Chemglass Life Sciences Supplier No. CG176105
Funnel, Vial, PTFE, 1.4" OD, 18-400 GPI Thread. Funnel machined from PTFE screws securely onto any standard laboratory vial having a GPI thread. Design allows for easy, safe and mess free transfer of product.