Learn More
Chemglass Life Sciences Funnel Only, 47mm, 300ml, Graduated
SDP
Catalog No. CG142002
Encompass_Preferred
Funnel Only, 47mm, 300ml, Graduated. Funnel for the CG142001 filtration support assembly. Glassware system for vacuum filtration using 47mm membranes. Assembly is designed for use with vacuum manifolds or optional plastic coated filtering flask. FUNNEL ONLY.
Catalog No. CG142002 Supplier Chemglass Life Sciences Supplier No. CG142002
Funnel Only, 47mm, 300ml, Graduated. Funnel for the CG142001 filtration support assembly. Glassware system for vacuum filtration using 47mm membranes. Assembly is designed for use with vacuum manifolds or optional plastic coated filtering flask. FUNNEL ONLY.