Learn More
MilliporeSigma FUNNELL/MILIFLX .45MICR/24/PK
Catalog No. NC9915450
Encompass
Presterilized funnel and filter unit Device Materials PE SAN Filter Brand Name Durapore Filter Material PVDF Product Use Broad chemical compatibility and low-protein-binding Filter Color white Filter Surface plain
Catalog No. NC9915450 Supplier MilliporeSigma Supplier No. MXHVWP124
Presterilized funnel and filter unit Device Materials PE SAN Filter Brand Name Durapore Filter Material PVDF Product Use Broad chemical compatibility and low-protein-binding Filter Color white Filter Surface plain