Learn More
Foxx Life Sciences Borosil Funnels with 60 Angle Bowls and 50mm Stems, Medium (M), 20/CS
Catalog No. 501542873
Encompass_Preferred
Borosil Short Stem Funnels are built for durability & long-term use. Funnels have 60 bowl angles for perfect fit with filter papers. Used for filtration & decanting of a wide variety of liquids. Built from Borosil's world-renowned 3.3 borosilicate, low-expansion, chemically resistant glass. Avail. in a variety of sizes from 25mm diameter (XS, Extra Small) to 100mm diameter (XXL, 2X-Extra Large). Borosil Short Stem Funnels comply with DIN/ISO 4798. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-154-2873 Supplier Foxx Life Sciences Supplier No. 6140065
Borosil Short Stem Funnels are built for durability & long-term use. Funnels have 60 bowl angles for perfect fit with filter papers. Used for filtration & decanting of a wide variety of liquids. Built from Borosil's world-renowned 3.3 borosilicate, low-expansion, chemically resistant glass. Avail. in a variety of sizes from 25mm diameter (XS, Extra Small) to 100mm diameter (XXL, 2X-Extra Large). Borosil Short Stem Funnels comply with DIN/ISO 4798. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity & reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free & incredibly resistant to thermal shock.