Learn More
Foxx Life Sciences Borosil® Funnels with 60 Angle Bowls and 65mm Stems, Large (L), 20/CS
Catalog No. 501720241
Encompass_Preferred
Borosil® Short Stem Funnels are built for durability & long-term use. Funnels have 60 bowl angles for perfect fit w/ filter papers. Used for filtration & decanting of a wide variety of liquids. Built from Borosil's world-renowned 3.3 borosilicate, low-expansion, chemically resistant glass. Available in a variety of sizes from 25mm diameter (XS, Extra Small) to 100mm diameter (XXL, 2X-Extra Large). Borosil® Short Stem Funnels comply w/ DIN/ISO 4798. In collaboration w/ Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.
Catalog No. 50-172-0241 Supplier Foxx Life Sciences Supplier No. 6140069
Borosil® Short Stem Funnels are built for durability & long-term use. Funnels have 60 bowl angles for perfect fit w/ filter papers. Used for filtration & decanting of a wide variety of liquids. Built from Borosil's world-renowned 3.3 borosilicate, low-expansion, chemically resistant glass. Available in a variety of sizes from 25mm diameter (XS, Extra Small) to 100mm diameter (XXL, 2X-Extra Large). Borosil® Short Stem Funnels comply w/ DIN/ISO 4798. In collaboration w/ Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, & reliability at competitive prices. & thanks to a unique annealing process, Borosil® Glassware is stress-free & incredibly resistant to thermal shock.