Learn More
Chemglass Life Sciences 60mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stpk, 265mm OAH
SDP
Catalog No. CG171312
Encompass_Preferred
60mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stopcock, 265mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171312 Supplier Chemglass Life Sciences Supplier No. CG171312
60mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stopcock, 265mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.