Learn More
Chemglass Life Sciences 1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm PTFE Metering Stpk, 525mm OAH
SDP
Catalog No. CG171320
Encompass_Preferred
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm PTFE Metering Stopcock, 525mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171320 Supplier Chemglass Life Sciences Supplier No. CG171320
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm PTFE Metering Stopcock, 525mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.