Learn More
Chemglass Life Sciences 50mm Powder Funnel, Modified, 14/20 Inner Joint, 15mm Flat Sided
SDP
Catalog No. CG172110
Encompass_Preferred
50mm Powder Funnel, Modified, 14/20 Inner Joint, 15mm Flat Sided. Similar to CG-1720, but with one side flattened to allow funnel to be used in multi-neck flasks where space is limited. Flat side dimension is the distance between the center of the standard taper inner joint and the flattened side of the funnel. Funnels have 60º angled sides with a standard taper inner joint.
Catalog No. CG172110 Supplier Chemglass Life Sciences Supplier No. CG172110
50mm Powder Funnel, Modified, 14/20 Inner Joint, 15mm Flat Sided. Similar to CG-1720, but with one side flattened to allow funnel to be used in multi-neck flasks where space is limited. Flat side dimension is the distance between the center of the standard taper inner joint and the flattened side of the funnel. Funnels have 60º angled sides with a standard taper inner joint.