Learn More
Chemglass Life Sciences 75mm Powder Funnel, 24/40 Inner Joint, 165mm OAL
SDP
Catalog No. CG1720L01
Encompass_Preferred
75mm Powder Funnel, 24/40 Inner Joint, 165mm OAL. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnel has steep sides to permit free-flow of solid materials. With standard taper inner joint.
Catalog No. CG1720L01 Supplier Chemglass Life Sciences Supplier No. CG1720L01
75mm Powder Funnel, 24/40 Inner Joint, 165mm OAL. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnel has steep sides to permit free-flow of solid materials. With standard taper inner joint.