Learn More
Chemglass Life Sciences 75mm Powder Funnel, 24/40 Inner Joint
SDP
Catalog No. CG172001
Encompass_Preferred
75mm Powder Funnel, 24/40 Inner Joint. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60º side angle and a standard taper inner joint.
Catalog No. CG172001 Supplier Chemglass Life Sciences Supplier No. CG172001
75mm Powder Funnel, 24/40 Inner Joint. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60º side angle and a standard taper inner joint.