Learn More
Chemglass Life Sciences 75mm Powder Funnel, Offset, 24/40 Inner Joint, C to C Offset is 30mm
SDP
Catalog No. CG172201
Encompass_Preferred
75mm Powder Funnel, Offset, 24/40 Inner Joint, C to C Offset is 30mm. Funnel has a lower stem made with an offset to permit use with multi-neck flasks.
Catalog No. CG172201 Supplier Chemglass Life Sciences Supplier No. CG172201
75mm Powder Funnel, Offset, 24/40 Inner Joint, C to C Offset is 30mm. Funnel has a lower stem made with an offset to permit use with multi-neck flasks.