Learn More
Medchemexpress LLC FUOM/Fucose Mutarota 50ug | 50ug
SDP
Catalog No. 5000186167
Encompass_Preferred
The FUOM/fucose mutarotase protein has a crucial interconversion effect on - and -L-fucose with -L-fucose being dominant (70 5%) The -form undergoes metabolic processes via the salvage pathway producing GDP-L-fucose that is transported to the endoplasmic reticulum FUOM/Fucose Mutarotase Protein Human (His) is the recombinant human-derived FUOM/Fucose Mutarotase protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-86167 Supplier Medchemexpress LLC Supplier No. HYP7694050UG
The FUOM/fucose mutarotase protein has a crucial interconversion effect on - and -L-fucose with -L-fucose being dominant (70 5%) The -form undergoes metabolic processes via the salvage pathway producing GDP-L-fucose that is transported to the endoplasmic reticulum FUOM/Fucose Mutarotase Protein Human (His) is the recombinant human-derived FUOM/Fucose Mutarotase protein expressed by E coli with N-His labeled tag