Learn More
AdipoGen FURA 2-AM (1 mg)
SDP
Supplier:  AdipoGen CDXF0014M001
Encompass_Preferred
FURA 2-AM (1 mg). CAS: 108964-32-5. Formula: C44H47N3O24. MW: 1001.87. Synthetic. The fluorescent probe FURA 2/AM is a membrane-permeable derivative of the calcium chelator FURA 2. Inside cells, FURA 2/AM is rapidly hydrolyzed by cytoplasmic esterases to FURA 2, which remains within the cell. Non-invasive, intracellular calcium indicator that is UV light-excitable. Fluorescent Ca2+ indicator, selective for Ca2+ over other divalent cations Mg2+, Zn2+, Fe2+ and Mn2+. Spectral properties in the presence of low calcium concentrations (lambdaex = 362nm, lambdaem = 512nm) differ from those during high calcium conditions (lambdaex = 335nm, lambdaem = 505nm). FURA 2 is preferred for ratio-imaging microscopy, in which it is more practical to change excitation wavelengths than emission wavelengths. Spectral Data: lambdaex 355nm; lambdaem 495nm in 0.1 M Tris pH 8.0.
Catalog No. 50-149-0379