Learn More
Abcam Fura-2 AM, Ca2+ selective fluorescent indicator, 10UNIT
Catalog No. 502584556
Encompass_Preferred
MW 1001.8 Da, Purity >95%. High affinity Ca2+ selective fluorescent indicator (Kd = 145 nM). Rapidly metabolized by cytoplasmic esterases leading to the active dye Fura 2. Cell-permeable. Excitation spectrum dependent on [Ca2+]i. Ex340/380/Em505.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4556 Supplier Abcam Supplier No. AB12087310UNIT
Add to Cart
Add to Cart
MW 1001.8 Da, Purity >95%. High affinity Ca2+ selective fluorescent indicator (Kd = 145 nM). Rapidly metabolized by cytoplasmic esterases leading to the active dye Fura 2. Cell-permeable. Excitation spectrum dependent on [Ca2+]i. Ex340/380/Em505.
The product is subject to the following: Abcam Restricted Use Statement