Learn More
Abcam Fura-2 AM, Ca2+ selective fluorescent indicator, 50UG
Catalog No. 502584553
Encompass_Preferred
MW 1001.8 Da, Purity >95%. High affinity Ca2+ selective fluorescent indicator (Kd = 145 nM). Rapidly metabolized by cytoplasmic esterases leading to the active dye Fura 2. Cell-permeable. Excitation spectrum dependent on [Ca2+]i. Ex340/380/Em505.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4553 Supplier Abcam Supplier No. AB12087350UG
MW 1001.8 Da, Purity >95%. High affinity Ca2+ selective fluorescent indicator (Kd = 145 nM). Rapidly metabolized by cytoplasmic esterases leading to the active dye Fura 2. Cell-permeable. Excitation spectrum dependent on [Ca2+]i. Ex340/380/Em505.
The product is subject to the following: Abcam Restricted Use Statement