Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fura 2-AM, MP Biomedicals™

Catalog No. MP021522865 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021522865 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021522865 Supplier MP Biomedicals, Inc Supplier No. 02152286.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Permeates cell membranes. Fura 2-AM, MP Biomedicals is a fluorescent probe for Ca2+ and is used to determine cystolic Ca2+. Fura 2-AM is the acetoxymethyl ester of Fura 2. It is metabolized in vivo to the active ligand Fura 2.

  • Able to permeate cell membranes and load Fura 2 into cells
  • Inside cells, Fura 2-AM is rapidly hydrolyzed by cytoplasmic esterases
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108964-32-5
Molecular Formula C44H47N3O24
Molecular Weight (g/mol) 1001.857
InChI Key VPSRLGDRGCKUTK-UHFFFAOYSA-N
Synonym fura 2-am, fura-2 pentakis acetoxymethyl ester, bis acetoxymethyl 2,2'-2-5-acetoxymethoxy carbonyl oxazol-2-yl-5-2-2-bis 2-acetoxymethoxy-2-oxoethyl amino-5-methylphenoxy ethoxy benzofuran-6-yl azanediyl diacetate, fura-2 am, fura-2-acetoxymethyl ester, 1-2-5-carboxyoxazol-2-yl-6-aminobenzofuran-5-oxy-2-2′-amino-5′-methylphenoxy-ethane-n,n,n′n′-tetraacetic acid pentaacetoxymethyl ester, 1-6-amino-2-5-carboxy-2-oxazolyl-5-benzofuranyloxy-2-2-amino-5-methylphenoxy ethane-n,n,n',n'-tetraacetic acid, pentaacetoxymethyl ester, fura-2/am, fura-2 acetoxymethyl ester, fura 2-am hplc
PubChem CID 3364574
IUPAC Name acetyloxymethyl 2-[6-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]-1-benzofuran-2-yl]-1,3-oxazole-5-carboxylate
SMILES CC1=CC(=C(C=C1)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)OCCOC2=C(C=C3C(=C2)C=C(O3)C4=NC=C(O4)C(=O)OCOC(=O)C)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C

Specifications

Quantity 500 μg
Solubility Information The acetoxymethyl ester of the chelator is soluble in dimethylsulfoxide (10mg/mL) and in nonpolar solvents such as chloroform, acetone and benzene. It is insoluble in water or methanol.
Formula Weight 1001.9
Physical Form Solid
Recommended Storage −20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.