Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fura 2-Am MP Biomedicals

Catalog No. ICN15228601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15228601 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15228601 Supplier MP Biomedicals, Inc Supplier No. 0215228601

Chemical Identifiers

CAS 108964-32-5
Molecular Formula C44H47N3O24
Molecular Weight (g/mol) 1001.857
InChI Key VPSRLGDRGCKUTK-UHFFFAOYSA-N
Synonym 1-[6-Amino-2-(5-carboxy-2-oxazolyl)-5-benzofuranyloxy]-2-(2-amino-5-methylphenoxy) ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester
PubChem CID 3364574
IUPAC Name acetyloxymethyl 2-[6-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-[2-[2-[bis[2-(acetyloxymethoxy)-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]-1-benzofuran-2-yl]-1,3-oxazole-5-carboxylate
SMILES CC1=CC(=C(C=C1)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)OCCOC2=C(C=C3C(=C2)C=C(O3)C4=NC=C(O4)C(=O)OCOC(=O)C)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C

Specifications

Formula Weight 1001.9
Quantity 1 mg
Chemical Name or Material Fura 2-Am
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.