Learn More
Cayman Chemical Fura-2 Leakage Resistant AM, Purity: ≥95%, 1 mg
SDP
Catalog No. 502911028
Encompass_Preferred
A cell-permeable, ratiometric fluorescent calcium indicator; binds to calcium (Kd = 145 nm) and displays excitation maxima of 340 and 380 nm when calcium-bound or calcium-free, respectively, when the emission is fixed at 505 nm
Catalog No. 50-291-1028 Supplier Cayman Chemical Supplier No. 349931 MG
A cell-permeable, ratiometric fluorescent calcium indicator; binds to calcium (Kd = 145 nm) and displays excitation maxima of 340 and 380 nm when calcium-bound or calcium-free, respectively, when the emission is fixed at 505 nm