Learn More
Aat Bioquest Fura-8 AM (1 mg)
SDP
Catalog No. 502249937
Encompass_Preferred
Although Fura-2 has become the preferred excitation-ratioable calcium indicator of choice it has certain limitations e.g. lower sensitivity than single wavelength calcium indicators such as Fluo-8 and Cal-520. This catalog is for 1 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9937 Supplier Aat Bioquest Supplier No. 21055
Although Fura-2 has become the preferred excitation-ratioable calcium indicator of choice it has certain limitations e.g. lower sensitivity than single wavelength calcium indicators such as Fluo-8 and Cal-520. This catalog is for 1 mg.