Learn More
Medchemexpress LLC Fura-FF AM | 348079-12-9 | 95.0% | 50 UG
SDP
Catalog No. 5000338297
Encompass_Preferred
Fura-FF AM is a cell-permeable acetoxymethyl ester of fura-FF, which is a fluorescent calcium indicator. It is hydrolyzed by intracellular esterases to release fura-FF within cells. Fura-FF is a difluorinated derivative of the calcium indicator fura-2. Compared to fura-2, fura-FF exhibits a low affinity for calcium, making it suitable for studying compartments with high concentrations of calcium.
  • Cell-permeable acetoxymethyl ester
  • Fluorescent calcium indicator
  • Hydrolyzed by intracellular esterases to release fura-FF within cells
  • Difluorinated derivative of fura-2
  • Low affinity for calcium compared to fura-2
  • Suitable for studying compartments with high concentrations of calcium
Catalog No. 50-003-38297 Supplier Medchemexpress LLC Supplier No. HY13057550UG
Fura-FF AM is a cell-permeable acetoxymethyl ester of fura-FF, which is a fluorescent calcium indicator. It is hydrolyzed by intracellular esterases to release fura-FF within cells. Fura-FF is a difluorinated derivative of the calcium indicator fura-2. Compared to fura-2, fura-FF exhibits a low affinity for calcium, making it suitable for studying compartments with high concentrations of calcium.
  • Cell-permeable acetoxymethyl ester
  • Fluorescent calcium indicator
  • Hydrolyzed by intracellular esterases to release fura-FF within cells
  • Difluorinated derivative of fura-2
  • Low affinity for calcium compared to fura-2
  • Suitable for studying compartments with high concentrations of calcium