Learn More
Aat Bioquest Fura Red AM CAS 149732-62-7 (10x50 ug)
SDP
Catalog No. 502249932
Encompass_Preferred
Fura Red is a visible light-excitable fura-2 analog that offers unique possibilities for ratiometric measurement of calcium ion in single cells by microphotometry imaging or flow cytometry when used with single excitation green-fluorescent calcium indicators. This catalog is for 10x50 ug.
This item is not returnable. View return policy
Catalog No. 50-224-9932 Supplier Aat Bioquest Supplier No. 21048
Fura Red is a visible light-excitable fura-2 analog that offers unique possibilities for ratiometric measurement of calcium ion in single cells by microphotometry imaging or flow cytometry when used with single excitation green-fluorescent calcium indicators. This catalog is for 10x50 ug.