Learn More
Medchemexpress LLC Fura Red AM | 218280-33-2 | 91.0% | 1088.99 | 1 MG
SDP
Catalog No. 5000430761
Encompass_Preferred
Fura Red AM is a fluorescent probe that can be used to visualize Ca²⁺ distribution in the cytoplasm of undifferentiated cells. It operates at an excitation wavelength of 488 nm.
  • Fluorescent probe for visualizing Ca²⁺ distribution
  • Suitable for use with undifferentiated cells
  • Operates at an excitation wavelength of 488 nm
Catalog No. 50-004-30761 Supplier Medchemexpress LLC Supplier No. HYD19031MG
Fura Red AM is a fluorescent probe that can be used to visualize Ca²⁺ distribution in the cytoplasm of undifferentiated cells. It operates at an excitation wavelength of 488 nm.
  • Fluorescent probe for visualizing Ca²⁺ distribution
  • Suitable for use with undifferentiated cells
  • Operates at an excitation wavelength of 488 nm