Learn More
Medchemexpress LLC HY-107204 5mg Medchemexpress, Furafylline CAS:80288-49-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1072045MG
Encompass_Preferred
Medchemexpress, HY-107204 5mg Furafylline CAS:80288-49-9 Furafylline is a potent and selective inhibitor of human cytochrome P450IA2 with an IC50 of 0.07 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8369