Learn More
Medchemexpress LLC Furafylline | 80288-49-9 | 260.25 g·mol⁻1 | C12H12N4O3 | 10 MG
SDP
Catalog No. 5000239865
Encompass_Preferred
Furafylline is a research-grade, selective inhibitor of human cytochrome P450 1A2 (CYP1A2) commonly used in in vitro studies of drug metabolism and enzyme mechanism. It is typically provided as a solid for dissolution in solvents such as DMSO and is intended for research use only.
  • Selective inhibitor of CYP1A2 with an IC50 of 0.07 μM.
  • Suitable for in vitro inhibition and drug-drug interaction studies.
  • Molecular formula C12H12N4O3; molecular weight 260.25 g·mol⁻1.
  • Powder storage at -20°C; in solvent storage at -80°C for long term.
  • Provided as an analytical-grade solid for research applications.
Catalog No. 50-002-39865 Supplier Medchemexpress LLC Supplier No. HY10720410MG
Furafylline is a research-grade, selective inhibitor of human cytochrome P450 1A2 (CYP1A2) commonly used in in vitro studies of drug metabolism and enzyme mechanism. It is typically provided as a solid for dissolution in solvents such as DMSO and is intended for research use only.
  • Selective inhibitor of CYP1A2 with an IC50 of 0.07 μM.
  • Suitable for in vitro inhibition and drug-drug interaction studies.
  • Molecular formula C12H12N4O3; molecular weight 260.25 g·mol⁻1.
  • Powder storage at -20°C; in solvent storage at -80°C for long term.
  • Provided as an analytical-grade solid for research applications.