Learn More
Abcam Furafylline, PDE inhibitor, 5MG
Catalog No. 502704960
Encompass_Preferred
MW 260.25 Da, Purity >98%. PDE inhibitor. Selective cytochrome P450IA2 inhibitor (IC₅₀ = 2.4 μM). Methyl xanthine derivative. Shows an expanded duration of action compared to theophylline (ab142599) in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4960 Supplier Abcam Supplier No. AB1460215MG
MW 260.25 Da, Purity >98%. PDE inhibitor. Selective cytochrome P450IA2 inhibitor (IC₅₀ = 2.4 μM). Methyl xanthine derivative. Shows an expanded duration of action compared to theophylline (ab142599) in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement