Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Furaltadone hydrochloride | 3759-92-0 | 99.46% | 1 ML
SDP

Catalog No. 5000411581
Encompass_Preferred

Furaltadone hydrochloride is an orally active antibacterial agent suitable for studying intestinal Salmonella infections. It demonstrates in vitro inhibitory effects on Staphylococcus and also shows inhibitory effects on allergic reactions. This product can be used for the study of intestinal Salmonella infections and suppresses IgE-mediated allergic response through the inhibition of Lyn/Syk pathway in mast cells.

  • Orally active antibacterial agent
  • Inhibitory effects on Staphylococcus in vitro
  • Inhibitory effects on allergic reactions
  • Solid appearance, light yellow to yellow color
  • Molecular Weight: 360.75
  • Formula: C13H17ClN4O6
  • Can be used for the study of intestinal Salmonella infections
  • Suppresses IgE-mediated allergic response through the inhibition of Lyn/Syk pathway in mast cells

Catalog No. 50-004-11581 Supplier Medchemexpress LLC Supplier No. HYB114810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Furaltadone hydrochloride is an orally active antibacterial agent suitable for studying intestinal Salmonella infections. It demonstrates in vitro inhibitory effects on Staphylococcus and also shows inhibitory effects on allergic reactions. This product can be used for the study of intestinal Salmonella infections and suppresses IgE-mediated allergic response through the inhibition of Lyn/Syk pathway in mast cells.

  • Orally active antibacterial agent
  • Inhibitory effects on Staphylococcus in vitro
  • Inhibitory effects on allergic reactions
  • Solid appearance, light yellow to yellow color
  • Molecular Weight: 360.75
  • Formula: C13H17ClN4O6
  • Can be used for the study of intestinal Salmonella infections
  • Suppresses IgE-mediated allergic response through the inhibition of Lyn/Syk pathway in mast cells

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.