Learn More
Medchemexpress LLC Furamidine dihydrochloride | 55368-40-6 | 99.0% | 377.27 | 10 MG
SDP
Catalog No. 5000343505
Encompass_Preferred
Furamidine dihydrochloride (DB75 dihydrochloride) is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor with an IC50 of 9.4 μM. It exhibits selectivity for PRMT1 over PRMT5, PRMT6, and PRMT4. This compound also acts as a potent, reversible, and competitive tyrosyl-DNA phosphodiesterase 1 (TDP-1) inhibitor, with slightly stronger inhibition observed with duplex DNA. It additionally functions as an antiparasite agent. Intended for research use only.
  • Selective PRMT1 inhibitor
  • Potent, reversible, and competitive TDP-1 inhibitor
  • Effective with both single- and double-stranded DNA substrates
  • Functions as an antiparasite agent
  • Inhibits cell growth in most leukemia cell lines
  • Significantly reduces expression level of methylated GFP-ALY protein
  • Can intercalate between GC base pairs of duplex DNA
Catalog No. 50-003-43505 Supplier Medchemexpress LLC Supplier No. HY11013710MG
Furamidine dihydrochloride (DB75 dihydrochloride) is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor with an IC50 of 9.4 μM. It exhibits selectivity for PRMT1 over PRMT5, PRMT6, and PRMT4. This compound also acts as a potent, reversible, and competitive tyrosyl-DNA phosphodiesterase 1 (TDP-1) inhibitor, with slightly stronger inhibition observed with duplex DNA. It additionally functions as an antiparasite agent. Intended for research use only.
  • Selective PRMT1 inhibitor
  • Potent, reversible, and competitive TDP-1 inhibitor
  • Effective with both single- and double-stranded DNA substrates
  • Functions as an antiparasite agent
  • Inhibits cell growth in most leukemia cell lines
  • Significantly reduces expression level of methylated GFP-ALY protein
  • Can intercalate between GC base pairs of duplex DNA