Learn More
Medchemexpress LLC Furamidine dihydrochloride | 55368-40-6 | 99.0% | 377.27 | 100 MG
SDP
Catalog No. 5000343504
Encompass_Preferred
Furamidine dihydrochloride is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor, exhibiting an IC50 of 9.4 μM. It demonstrates selectivity for PRMT1 over PRMT5, PRMT6, and PRMT4. Additionally, it functions as a potent, reversible, and competitive tyrosyl-DNA phosphodiesterase 1 (TDP-1) inhibitor, effective with both single- and double-stranded DNA substrates. This compound also possesses antiparasite agent properties.
  • Selective PRMT1 inhibitor
  • Potent, reversible, and competitive TDP-1 inhibitor
  • Effective against single- and double-stranded DNA substrates
  • Acts as an antiparasite agent
  • Inhibits cell growth in most leukemia cell lines (except HEL cells with JAK2V617F mutations)
  • Reduces expression of methylated GFP-ALY protein in 293T cells
  • Can intercalate between GC base pairs of duplex DNA
Catalog No. 50-003-43504 Supplier Medchemexpress LLC Supplier No. HY110137100MG
Furamidine dihydrochloride is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor, exhibiting an IC50 of 9.4 μM. It demonstrates selectivity for PRMT1 over PRMT5, PRMT6, and PRMT4. Additionally, it functions as a potent, reversible, and competitive tyrosyl-DNA phosphodiesterase 1 (TDP-1) inhibitor, effective with both single- and double-stranded DNA substrates. This compound also possesses antiparasite agent properties.
  • Selective PRMT1 inhibitor
  • Potent, reversible, and competitive TDP-1 inhibitor
  • Effective against single- and double-stranded DNA substrates
  • Acts as an antiparasite agent
  • Inhibits cell growth in most leukemia cell lines (except HEL cells with JAK2V617F mutations)
  • Reduces expression of methylated GFP-ALY protein in 293T cells
  • Can intercalate between GC base pairs of duplex DNA