Learn More
Abcam Furamidine dihydrochloride, 25MG
Catalog No. 502671202
Encompass_Preferred
MW 304.3 Da, Purity >=98%. A potent, selective and cell-permeable protein arginine methyltransferase 1 (PRMT1) inhibitor (IC₅₀ = 9.4 μM). Displays selectivity over PRMT5, PRMT6 and CARM1 (IC₅₀ values are 166, 283 and >400 μM respectively). Inhibits cell proliferation in leukemia cell lines.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1202 Supplier Abcam Supplier No. AB28709825MG
MW 304.3 Da, Purity >=98%. A potent, selective and cell-permeable protein arginine methyltransferase 1 (PRMT1) inhibitor (IC₅₀ = 9.4 μM). Displays selectivity over PRMT5, PRMT6 and CARM1 (IC₅₀ values are 166, 283 and >400 μM respectively). Inhibits cell proliferation in leukemia cell lines.
The product is subject to the following: Abcam Restricted Use Statement