Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Furan

Catalog No. 501854216
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-4216 500 mL
50-185-4215 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-4216 Supplier Sigma Aldrich Supplier No. 185922500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Furan is a five membered heterocyclic compound that is highly volatile, flammable and a potential carcinogen. it undergoes diels-alder reaction with 2-bromoacrolein catalyzed by oxazaborolidine to yield chiral 7-oxabicyclo[2.2.1]heptene derivatives. cationic bis(4-tert-butyloxazoline)cu(ii) complex catalyzed enantioselective diels-alder reaction between acryloyl oxazolidinone and furan to afford ent-shikimic acid has been described. lewis acid catalyzed diels-alder reaction between furan and some dienophiles affords substituted cyclohexenols and cyclohexadienols.furan was used in the following processes:
  • Preparation of the starting material required for the synthesis of calix[6]pyrrole
  • To investigate the kinetics and mechanism of reactions of chlorine atoms with volatile organic compounds
  • Catalytic transformation of furan to aromatics and olefins
TRUSTED_SUSTAINABILITY

Specifications

CAS 110-00-9
Density 0.936 g/mL (at 25°C (literature))
Boiling Point 32°C (758 mmHg)
Molecular Formula C4H4O
Refractive Index n20/D 1.421 (literature)
Linear Formula C4H4O
MDL Number MFCD00003222
Quantity 500 mL
Molecular Weight (g/mol) 68.07
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.