Learn More
GLPBIO TECHNOLOGY INC Furan Fatty Acid F5 (Synonyms: Furan Fatty Acid F5) 1mg
Catalog No. NC3781151
Encompass
Furan fatty acid F5 is a furan fatty acid originally isolated from northern pike (E. lucius). Levels of furan fatty acid F5 are increased in the liver of starving cod.
Catalog No. NC3781151 Supplier GLPBIO TECHNOLOGY INC Supplier No. GC18801
Furan fatty acid F5 is a furan fatty acid originally isolated from northern pike (E. lucius). Levels of furan fatty acid F5 are increased in the liver of starving cod.