Learn More
AdipoGen Furanone C-30
SDP
Supplier:  AdipoGen CDXB0220M100
Encompass_Preferred
Chemical. CAS 247167-54-0. Formula C5H2Br2O2. MW 253.88. Synthetic Quorum sensing inhibitor. Interfers with the actions of acyl-HSL molecules and inhibits gene expression regulated by quorum-sensing systems.
Catalog No. 50-256-1638