Learn More
AdipoGen Furanone C-30
SDP
Catalog No. 502561640
Encompass_Preferred
Chemical. CAS 247167-54-0. Formula C5H2Br2O2. MW 253.88. Synthetic Quorum sensing inhibitor. Interfers with the actions of acyl-HSL molecules and inhibits gene expression regulated by quorum-sensing systems.
Catalog No. 50-256-1640 Supplier AdipoGen Supplier No. CDXB0220M010
Add to Cart
Add to Cart
Chemical. CAS 247167-54-0. Formula C5H2Br2O2. MW 253.88. Synthetic Quorum sensing inhibitor. Interfers with the actions of acyl-HSL molecules and inhibits gene expression regulated by quorum-sensing systems.